missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium acetate, ACS, 99.0-102.0%

Manufacturer:  Alfa Aesar™ 1219830

 View more versions of this product

Catalog No. AA1219830



Specifications

Specifications

Barium acetate
C4H6BaO4
UN1564
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
CC(=O)[O-].CC(=O)[O-].[Ba+2]
255.415
255.43
99.0 to 102.0%
2.47g/mL
250g
543-80-6
MFCD00012447
14,965
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
barium(2+);diacetate
10980
Crystalline
ACS
Odorless

Chemical Identifiers

543-80-6
255.415
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
10980
CC(=O)[O-].CC(=O)[O-].[Ba+2]
C4H6BaO4
MFCD00012447
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
barium(2+);diacetate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Documents