missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Barium Acetate, 99.0 to 102.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent, 99%

$98.90 - $98.90

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.415
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Ba+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045624 View Documents Honeywell Fluka™
243671100G
100g Poly Bottle Each for $98.90
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

543-80-6
255.415
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
10980
CC(=O)[O-].CC(=O)[O-].[Ba+2]
C4H6BaO4
MFCD00012447
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
barium(2+);diacetate

Specifications

543-80-6
(CH3COO)2Ba
NONH for all modes of transport
Powder or crystals/granules
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
barium(2+);diacetate
10980
99.0 to 102.0%
2.46g/cm3
White
Barium Acetate
C4H6BaO4
MFCD00012447
3693411
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
CC(=O)[O-].CC(=O)[O-].[Ba+2]
255.415
255.42g/mol
ACS Reagent
Poly Bottle
100g
Safety and Handling

Safety and Handling

P261-P301 + P312 + P330

H302 + H332

EINECSNumber : 208-849-0

RTECSNumber : AF4550000