Please login to your online account to display your discounted pricing

Barium, 99+%, rods, ∽2cm diameter, in mineral oil, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC317865000

 View more versions of this product

Catalog No. AC317865000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Rod
Assay ≥99.0% Ba (incl. Sr)
Formula Weight 137.34
Quantity 500g
Color Gray
Merck Index 15, 960
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material Barium
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula Ba
Linear Formula Ba
MDL Number MFCD00134031
CAS 7440-39-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
In contact with water releases flammable gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Strontium (Sr) 0.8% max.

Specifications

Chemical Name or Material Barium
Name Note 99+%
CAS 7440-39-3
CAS Min % 99.0
CAS Max % 100.0
Physical Form Rod
Assay Percent Range 99+%
Molecular Formula Ba
Linear Formula Ba
Formula Weight 137.34
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
In contact with water releases flammable gas.
Reacts violently with water.

SDS & Safety Documents