Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium 2-methoxyethoxide, 99.5% (metals basis), 10% w/v in 2-methoxyethanol

Chemical Identifiers

Chemical Name or Material Barium 2-methoxyethoxide
CAS 115503-13-4
Linear Formula Ba(OCH2CH2OCH3)2
Formula Weight 287.5
MDL Number MFCD00673672
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41256AC

 
alfa aesar™
41256AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41256AE

 
alfa aesar™
41256AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 287.5
Chemical Name or Material Barium 2-methoxyethoxide
Physical Form Liquid
Assay 99.5%
Assay Percent Notes (metals basis)
Linear Formula Ba(OCH2CH2OCH3)2
MDL Number MFCD00673672
Sensitivity Moisture Sensitive
CAS 115503-13-4
UN Number UN3274
Health Hazard 1 Danger
Color Yellow
Concentration 10% w/v in 2-methoxyethanol
Quantity 25mL
Health Hazard 2 GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.