missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Barium 2-ethylhexanoate in 2-ethylhexanoic acid, Ba ≈17.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04484714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium 2-ethylhexanoate in 2-ethylhexanoic acid | |
Yellow | |
MFCD00058696 | |
barium 2-ethylhexanoate, barium bis 2-ethylhexanoate, barium 2+ ; 2-ethylhexanoate, hexanoic acid, 2-ethyl-, barium salt 2:1, acmc-20alrk, hexanoic acid, 2-ethyl-, barium salt, dsstox_cid_24888, dsstox_rid_80561, dsstox_gsid_44888, bis 2-ethylhexanoic acid barium salt | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ba+2] | |
423.74 | |
423.76 | |
Viscous Liquid |
2457-01-4 | |
C16H30BaO4 | |
25 g | |
VJFFDDQGMMQGTQ-UHFFFAOYNA-L | |
barium(2+);2-ethylhexanoate | |
102821 | |
Ba ≈17.5% |
Chemical Identifiers
2457-01-4 | |
423.74 | |
VJFFDDQGMMQGTQ-UHFFFAOYNA-L | |
102821 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ba+2] |
C16H30BaO4 | |
MFCD00058696 | |
barium 2-ethylhexanoate, barium bis 2-ethylhexanoate, barium 2+ ; 2-ethylhexanoate, hexanoic acid, 2-ethyl-, barium salt 2:1, acmc-20alrk, hexanoic acid, 2-ethyl-, barium salt, dsstox_cid_24888, dsstox_rid_80561, dsstox_gsid_44888, bis 2-ethylhexanoic acid barium salt | |
barium(2+);2-ethylhexanoate |
Safety and Handling
GHS H Statement
H331-H361-H302
Toxic if inhaled.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H361d
EINECSNumber : 219-535-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only