Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol

Chemical Identifiers

Chemical Name or Material Barium 1-methoxy-2-propoxide
CAS 130812-47-4
Linear Formula Ba[OCH(CH3)CH2OCH3]2
Formula Weight 315.55
MDL Number MFCD00078021
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4125706

  SDS
alfa aesar™
4125706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4125722

  SDS
alfa aesar™
4125722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 315.55
Chemical Name or Material Barium 1-methoxy-2-propoxide
Physical Form Liquid
Linear Formula Ba[OCH(CH3)CH2OCH3]2
MDL Number MFCD00078021
Sensitivity Air & Moisture Sensitive
CAS 130812-47-4
UN Number UN2924
Density 1.04g/mL
Health Hazard 1 Danger
Concentration 25% w/w in 1-methoxy-2-propanol
Quantity 5g
Flash Point 35° (95°F)
Health Hazard 2 GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.