missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bare Copper Wire
For general lab use and low voltage hook-ups.
Manufacturer: Fisher Scientific F18(EMD)
Description
Applications:
- As a fastener
- For electrodes
- To connect circuits
- As a source of copper in electrochemistry experiments
Order Info
Comes in 1 lb. spool (454g).
Specifications
Electrodes | |
General lab use and low voltage hook-ups, as a fastener, electrodes, to connect circuits, as a source of copper in electrochemistry experiments | |
199 ft. | |
0.0403 in. | |
1 lb. |
Bare Copper Wire | |
18 ga | |
60.65 m | |
0.102 cm | |
0.45 kg |