Upchurch Scientific™ Barbed Union Connectors
$12.96 - $12.96
Specifications
Type Barbed Union Connectors
For Use With (Equipment) Peristaltic Tubing
Material Polypropylene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
05-700-194 Upchurch Scientific™
UPP801X*
Connects 0.0625 in. I.D. tubing to 0.0625 in. I.D. tubing Pack of 10 Each for $12.96
Specifications Description & Specifications
Specifications
Barbed Union Connectors
Peristaltic Tubing
Polypropylene