Upchurch Scientific™ Barbed-to-Threaded Adapters
$9.50 - $9.94
Specifications
Type Barbed-to-Threaded Adapter
For Use With (Application) Connecting soft tubing to standard 1/4-28 internally-threaded ports
Material ETFE
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
29-008-1375 Upchurch Scientific™
UPP648
4.76mm Each for $9.50
29-008-412 Upchurch Scientific™
UPP646
1.57mm Each for $9.94
Specifications Description & Specifications
Specifications
Barbed-to-Threaded Adapter
Connecting soft tubing to standard 1/4-28 internally-threaded ports
ETFE