Upchurch Scientific™ Barbed Reducers
$14.30 - $14.30
Specifications
Type Barbed Reducing Adapter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
05-700-203 Upchurch Scientific™
UPP810X
To connect 3/8 in. ID (9.55 mm) Tubing to 1/4 in. ID (6.35 mm) Tubing Pack of 10 Each for $14.30
Specifications Description & Specifications
Specifications
Barbed Reducing Adapter