missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Banoxantrone dihydrochloride

Prodrug topoisomerase II inhibitor

$244.81 - $997.23

Specifications

Product Type Banoxantrone dihydrochloride
Inhibitors Banoxantrone dihydrochloride
Molecular Weight 517.4
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
4219/10 Tocris™
RDS421910
10mg Each for $244.81
4219/50 Tocris™
RDS421950
50mg Each for $997.23
Specifications

Description

Bioreductive prodrug; activated by hypoxia to yield the topoisomerase II inhibitor AQ4. Displays little to no cytotoxic activity against 60 tumor cell lines tested under oxic conditions (IC50 >100μM). Exhibits anti-tumor efficacy in vivo when combined with oxic cell cytotoxins; enhances the anti-tumor effect of cyclophosphamide (Cat. No. 4091) in a mouse tumor model.
  • Chemical Name: 1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione dihydrochloride
  • Formula: C22H28N4O6.2HCl
  • CAS Number: 252979-56-9
  • Solubility: Soluble to 50mM in water and to 25mM in DMSO)
  • Purity: >98
Description & Specifications

Specifications

Banoxantrone dihydrochloride
Banoxantrone dihydrochloride
517.4
>98