missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ BAN ORL 24

Potent and selective NOP antagonist

$250.58 - $1,042.86

Specifications

Product Type BAN ORL 24
Inhibitors BAN ORL 24
Molecular Weight 506.51
Purity >97
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
366110R Tocris™
366110
10mg Each for $250.58
366150R Tocris™
366150
50mg Each for $1,042.86
Specifications

Description

Potent and selective NOP receptor antagonist (IC50 values are 0.27, 2500, 6700 and > 10000nM for NOP, κ-,μ- and δ-receptors respectively). Inhibits nociceptin-induced stimulation of [35S]-GTPγS binding and Ca2+ mobilization in CHO cells in vitro and antagonizes NOP agonist-induced reduction in locomotor activity in vivo. Brain penetrant.
  • Chemical Name: (2R)-1-(Phenylmethyl)-N-[3-(spiro[isobenzofuran-1(3H),4'-piperidin]-1-yl)propyl-2-pyrrolidinecarboxamide
  • Formula: C27H35N3O2.2HCl
  • CAS Number: 1401463-54-4
  • Solubility: Soluble to 100mM in water and to 100mM in DMSO
  • Purity: >97
Description & Specifications

Specifications

BAN ORL 24
BAN ORL 24
506.51
>97