missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BAM 7, Tocris™

Selective Bax activator; induces Bax-mediated apoptosis

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C21H19N5O2S
Formula Weight 405.47
Synonym 3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4, 5-dione 4-[2-(2-Ethoxyphenyl)hydrazone]
Chemical Name or Material BAM 7
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
481010 R & D Systems
RDS481010
10mg Each for $244.81
481050 R & D Systems
RDS481050
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>99%
405.47
BAM 7
C21H19N5O2S
3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4, 5-dione 4-[2-(2-Ethoxyphenyl)hydrazone]

Specifications

10mg
>99%
405.47
BAM 7
331244-89-4
C21H19N5O2S
3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4, 5-dione 4-[2-(2-Ethoxyphenyl)hydrazone]

Safety and Handling

Recommended Storage : Store at +4°C