Please login to your online account to display your discounted pricing

Alfa Aesar™ BAM-12P

For cellular and molecular biology applications

Specifications

Product Type BAM-12P
Molecular Weight 1424.64
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-Gly-Arg-Pro-Glu
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66932LB0

 
alfa aesar™
J66932LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66932MA

 
alfa aesar™
J66932MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type BAM-12P
Molecular Weight 1424.64
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-Gly-Arg-Pro-Glu
Common Name Bovine Adrenal Medulla Dodecapeptide
Regulatory Status RUO
Purification Method HPLC