Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
Manufacturer:  Bal Supply 20112WS
 View more versions of this product
Catalog No. 05-885C
Description
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
  • 05-885C and 05-885E through K have screw-locking device
  • Work well on both O-ring and ball-and-socket joints
  • Others are intended for ball-and-socket joints only
Specifications
Specifications
Without screw locking device
Stainless Steel
Pinch Clamp
For Ball to Socket Joint Size: 12/1 and 12/5
12/Pk.