Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
Manufacturer: Bal Supply 20150WS
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
- 05-885C and 05-885E through K have screw-locking device
- Work well on both O-ring and ball-and-socket joints
- Others are intended for ball-and-socket joints only
Specifications
Screw locking device | |
Stainless Steel | |
Pinch Clamp |
For 50/30 Joints | |
6/Pk. |