Bal Supply Morton™ Stainless-Steel Culture Tube Closures
Ideal for use with standard glass culture tubes
Manufacturer:  Bal Supply 18144CL
 View more versions of this product
Catalog No. 05888C
Description
Description
Ideal for use with standard glass culture tubes
  • Non-contaminating, noncorrosive stainless-steel cylindrical closures
  • Top surface indentations allow adequate gas exchange under aerobic and anaerobic conditions, yet prevent buildup of dust and microorganisms
  • Reduce amount of specimen evaporation by 50% as compared to cotton plugging
  • Withstand repeated autoclaving without loss of resiliency
  • Available as plain or lanced closures
  • Lanced closures feature three resilient fingers that adjust to nominal variations in tube O.D
Specifications
Specifications
Standard glass culture tubes for microbiology and related applications
Autoclavable
Silver
3/4 in.
Lanced Closure
Stainless Steel
18 mm
18 mm