Please login to your online account to display your discounted pricing

Baker-flex™, Silica Gel IB2-F, J.T.Baker™

$395.36 - $614.48

Chemical Identifiers

Chemical Name or Material Silica Gel IB2-F
CAS 7631-86-9
Formula Weight 60.09
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

02003910

 
avantor performance
444904
Each for $614.48

02003909

 
avantor performance
444902
Each for $395.36
Description & Specifications

Specifications

Physical Form Crystal or Powder
Formula Weight 60.09
Quantity 25 Sheets (20 x 20cm) 8/case
Chemical Name or Material Silica Gel IB2-F
Odor Odorless
CAS 7631-86-9
Solubility Information Insoluble in water
Boiling Point 2230°C
Color White
Melting Point 1710°C

Flexible TLC Sheet