missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Avantor™ Baker-flex™, Silica Gel IB, J.T.Baker™

Flexible TLC Sheet

$420.11 - $692.91

Specifications

Chemical Name or Material Silica Gel IB
CAS 7631-86-9
View More Specs

Products 3
Catalog Number Mfr. No. Size (Metric) Price Quantity  
Catalog Number Mfr. No. Size (Metric) Price Quantity  
2003902 View Documents Avantor™
446200
Each for $431.05
 
2003903 View Documents Avantor™
446202
Each for $420.11
 
2003904 View Documents Avantor™
446204
Each for $692.91
 
Description

Description

Flexible TLC Sheet

Specifications

Specifications

Silica Gel IB
7631-86-9