Please login to your online account to display your discounted pricing

Baker-flex™, Cellulose PEI, J.T.Baker™


Flexible TLC Sheet

Manufacturer: avantor performance  447304

 View more versions of this product

Catalog No. 02002563

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystal or Powder
Chemical Name or Material Cellulose PEI
CAS 9004-34-6
Odor Odorless
Color White
Solubility Information Insoluble in water
Quantity 25 Sheets (20 x 20cm)

Specifications

Chemical Name or Material Cellulose PEI
CAS 9004-34-6