Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bak Ab-2, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Bak Ab-2, Rabbit Polyclonal Antibody.

$532.04 - $1,758.20

Specifications

Antigen Bak Ab-2
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Hamster
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1520P

 
thermo scientific™ lab vision™
RB1520P
Purified Ab with BSA and Azide 1mL Each for $1,600.00

RB1520P0

 
thermo scientific
RB1520P0
Purified Ab with BSA and Azide 100µL Each for $532.04

RB1520P1

 
thermo scientific™ lab vision™
RB1520P1
Purified Ab with BSA and Azide 500µL Each for $1,118.55

RB1520P1ABX

 
thermo scientific™ lab vision™
RB1520P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $1,221.39

RB1520PABX

 
thermo scientific™ lab vision™
RB1520PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,758.20

RB1520R7

 
thermo scientific™ lab vision™
RB1520R7
Ready-to-use for IHC (UVLP) 7mL Each for $966.39
Description & Specifications

Specifications

Antigen Bak Ab-2
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Hamster
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A synthetic peptide from the N-terminus of human Bak protein
Isotype IgG

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Bak Ab-2, Rabbit Polyclonal Antibody.

Bak or Bcl-2-homologous antagonist is a member of the Bcl-2 family of holologous proteins which function in the regulation of apoptosis. The Bcl-2 related proteins interact with one another through the formation of homo- and heterodimers. The susceptibility of cells to apoptotic stimuli is thought to be controlled by the relative ratios of the different Bcl-2 family proteins. Bak is a 211 aa protein which has been demonstrated to accelerate the rate of apoptosis in growth factor deprived murine lymphoid, neuronal and fibroblastic cell lines. Bak has been localized by immunohistochemistry to the cytosol of cells and stains in a punctate pattern characteristic of other bcl-2 family proteins, which have been localized to intracellular organelles.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Hamster (Weakly). Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human Bak protein

Molecular Weight: ∽24kDa

Positive Control: A431 cells, Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO