missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bak Ab-2, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Bak Ab-2, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ RB1520PABX

 View more versions of this product

Catalog No. RB1520PABX


 

Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Bak Ab-2, Rabbit Polyclonal Antibody.

Bak or Bcl-2-homologous antagonist is a member of the Bcl-2 family of holologous proteins which function in the regulation of apoptosis. The Bcl-2 related proteins interact with one another through the formation of homo- and heterodimers. The susceptibility of cells to apoptotic stimuli is thought to be controlled by the relative ratios of the different Bcl-2 family proteins. Bak is a 211 aa protein which has been demonstrated to accelerate the rate of apoptosis in growth factor deprived murine lymphoid, neuronal and fibroblastic cell lines. Bak has been localized by immunohistochemistry to the cytosol of cells and stains in a punctate pattern characteristic of other bcl-2 family proteins, which have been localized to intracellular organelles.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Hamster (Weakly). Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human Bak protein

Molecular Weight: ∽24kDa

Positive Control: A431 cells, Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Bak Ab-2
Polyclonal
Unlabeled
Concentrated
A synthetic peptide from the N-terminus of human Bak protein
A431 cells, tonsil
RUO
Primary
Hamster,Human
Western Blot
1mg/mL
Ab without BSA/Azide (Frozen)
Rabbit
IgG
1mL
Apoptosis
Polyclonal