Please login to your online account to display your discounted pricing

Alfa Aesar™ Bafilomycin A1

Specific, potent inhibitor of vacuolar ATPases

$263.22 - $263.22

Chemical Identifiers

Chemical Name or Material Bafilomycin A1
CAS 88899-55-2
Formula Weight 622.83
MDL Number MFCD06795130
Beilstein 4730700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61835MCR

  SDS
alfa aesar™
J61835MCR
1mg Each for $263.22
Description & Specifications

Specifications

Formula Weight 622.83
Chemical Name or Material Bafilomycin A1
Physical Form Oil or Film
CAS 88899-55-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD06795130
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1mg
Beilstein 4730700
Color Colorless
Storage Note 1 -30° to -10°C
RTECS Number RN9781000

Solubility
Soluble in methanol, ethanol, acetone and chloroform. Insoluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.