Please login to your online account to display your discounted pricing

BADGE, Tocris

PPARγ antagonist

$73.75 - $252.39

Chemical Identifiers

Chemical Name or Material BADGE
Assay Percent Range >90%
Molecular Formula C21H24O4
Formula Weight 340.42
Synonym 2, 2-Bis[4-(2, 3-epoxypropoxy)phenyl]propane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

132610

 
r & d systems
RDS132610
10mg Each for $73.75

132650

 
r & d systems
RDS132650
50mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 340.42
Quantity 10mg
Chemical Name or Material BADGE
Molecular Formula C21H24O4
CAS 1675-54-3
Recommended Storage Desiccate at -20°C
Assay Percent Range >90%
Synonym 2, 2-Bis[4-(2, 3-epoxypropoxy)phenyl]propane
Safety and Handling

Desiccate at -20°C