Please login to your online account to display your discounted pricing

baculovirus envelope gp64 protein Mouse, Clone: AcV5, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen baculovirus envelope gp64 protein
Clone AcV5
Host Species Mouse
Gene Alias BV gp64
Applications Western Blotting
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122721

 
affymetrix
14699581
50μg Each for $187.05

5013576

 
affymetrix
14699582
100μg Each for $390.05

501128901

 
affymetrix
14699585
500μg Each for $433.55
Description & Specifications

Specifications

Antigen baculovirus envelope gp64 protein
Clone AcV5
Host Species Mouse
Gene Alias BV gp64
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The AcV5 antibody reacts with the gp64 envelope protein of the baculovirus Autographa californica (AcMNPV).