Please login to your online account to display your discounted pricing

Alfa Aesar™ Bactenecin, Bovine


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66855LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66855LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Format Lyophilized powder
Sequence Arg-Leu-Cys-Arg-Ile-Val-Val-Ile-Arg-Val-Cys-Arg (Disulfide bridge: Cys3-Cys11)
Molecular Weight 1483.89
Storage Requirements -30° to -10°C
Quantity 5mg
Product Type Bactenecin, bovine
Cross Reactivity Bovine
Purification Method HPLC
Regulatory Status RUO