Please login to your online account to display your discounted pricing

Alfa Aesar™ Bactenecin, Bovine

For cellular and molecular biology applications

Specifications

Product Type Bactenecin, bovine
Molecular Weight 1483.89
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Arg-Leu-Cys-Arg-Ile-Val-Val-Ile-Arg-Val-Cys-Arg (Disulfide bridge: Cys3-Cys11)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66855LB0

 
alfa aesar™
J66855LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66855MA

 
alfa aesar™
J66855MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66855MB

 
alfa aesar™
J66855MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Bactenecin, bovine
Molecular Weight 1483.89
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Arg-Leu-Cys-Arg-Ile-Val-Val-Ile-Arg-Val-Cys-Arg (Disulfide bridge: Cys3-Cys11)
Regulatory Status RUO
Purification Method HPLC