missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ BacMam LRRK2-GFP Wild Type Reagent DFS Item

BacMam LRRK2-GFP Wild Type Reagent

Supplier:  Invitrogen™ A14170

Catalog No. A14170



Description

Description

BacMam LRRK2-GFP Wild Type Reagent is a modified insect cell virus (baculovirus) that enables easy, efficient, and titratable overexpression of full-length wild type LRRK2-GFP in your choice of cellular background, including primary neurons and neuroblastoma cell lines.

BacMam LRRK2-GFP Wild Type Reagent Lets You:

  • Overexpress full-length LRRK2-GFP in most cellular backgrounds
  • Visualize LRRK2 expression
  • Conduct the LRRK2 cellular assays you need
  • Understand the effects of therapeutically relevant LRRK2 mutations

    Use BacMam Technology with GFP for Expression and Visualization of LRRK2
    GFP (Green Fluorescent Protein) allows you to visually confirm LRRK2-GFP protein expression and compare localization of wild type with pathological mutants in various cellular backgrounds, especially primary neurons, while BacMam technology utilizes a modified baculovirus to deliver and express genes in mammalian cells with minimal effort and toxicity. BacMam enables efficient, titratable LRRK2-GFP protein expression in various mammalian cellular backgrounds including cell lines that may not be compatible with traditional transfection methods, such as primary neurons and neuroblastoma cell lines.

    Expand Your Battery of Cellular Assays for LRRK2
    Once full-length LRRK2-GFP has been introduced into your cellular background, the transduced cells allow you the flexibility to conduct many types of LRRK2 experiments, including studies of regulation of physiologically functional pathways (including signaling pathways leading to function) and identification of potential physiological substrates for LRRK2.

    Examine LRRK2 Mutations with BacMam Reagents
    Use therapeutic mutant reagents G2019S (Cat No A14174) along with the wild type LRRK2-GFP reagent to enable your research into LRRK2 mutations, including exploration of differences in cellular response to stimulus among different mutations.

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:
  • LRRK2 tools for advancing Parkinson's disease research
  • Kinase protein portfolio
  • LanthaScreen™ Eu Kinase Binding Assay
  • LanthaScreen™ Activity Assay
  • Learn More About BacMam Technology
  • Specifications

    Specifications

    Recombinant baculovirus particles suspended in Gibco™ Sf-900™ III SFM media supplemented with 3% heat-inactivated fetal bovine serum
    15 mL
    Store at 4°C. Do Not Freeze. Keep Sterile. Protect from light. Aliquot upon arrival.
    Overexpression of full-length wild type LRRK2-GFP in most cellular backgrounds
    SDS
    Documents

    Documents

    Promotions

    Promotions

    For Research Use Only. Not for use in diagnostic procedures.