Please login to your online account to display your discounted pricing

Alfa Aesar™ Bacitracin, 10mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Bacitracin
CAS 1405-87-4
Name Note 10mg/mL in distilled water, sterile-filtered
MDL Number MFCD00062640
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67048AC

  SDS
alfa aesar™
J67048AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bacitracin
MDL Number MFCD00062640
CAS 1405-87-4
TSCA TSCA
Name Note 10mg/mL in distilled water, sterile-filtered
Physical Form Liquid
Storage Note 1 -30° to -10°C
Quantity 25mL
RTECS Number CP0175000
Safety and Handling

-30° to -10°C