missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

B7-H4 Rat anti-Mouse, PE, Clone: Clone 9, eBioscience™

Rat Monoclonal Antibody

$170.28 - $473.88

Specifications

Antigen B7-H4
Clone Clone 9
Host Species Rat
Gene Alias B7H4, B7S1, B7-S1, B7X
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011030 Life Technologies
LS12597081
50μg Each for $170.28
5011031 Life Technologies
LS12597082
100μg Each for $302.28
5011032 Life Technologies
LS12597083
200μg Each for $473.88
Specifications

Description

The Clone 9 monoclonal antibody reacts with mouse B7S1, also known as B7H4. B7S1 is a newly discovered member of the B7 family. It is speculated that the costimulatory regulation of T cells by B7S1 is influenced by the activation status of B cells. The ligand for B7S1 has not been identified yet. This antibody is not blocking. Simultaneous double staining of cells with two anti-mouse B7-H4 antibodies, Clone 9 and 188, suggests that epitopes recognized by these mAbs are different and/or there is no steric hindrance when antibodies are used together.
Description & Specifications

Specifications

B7-H4
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
Clone 9
B7H4, B7S1, B7-S1, B7X
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer