Please login to your online account to display your discounted pricing

B7-H4 Mouse anti-Human, APC, Clone: H74, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen B7-H4
Clone H74
Host Species Mouse
Gene Alias B7H4, B7S1, B7-S1, B7X
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123167

 
life technologies
LS17594941
25 tests Each for $172.55

501129090

 
life technologies
LS17594942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen B7-H4
Clone H74
Host Species Mouse
Gene Alias B7H4, B7S1, B7-S1, B7X
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The H74 monoclonal antibody was generated and reacts with human B7-S1, also known as B7-H4 and B7x. Cross reactivity of this antibody to other proteins has not been determined. B7S1 is a newly discovered member of the B7 family. It is speculated that the costimulatory regulation of T cells by B7S1 is influenced by the activation status of B cells. While it is reported that BTLA is a counter receptor for B7S1, further studies are needed to definitely determine the B7-H4 ligand. H74 stains human B7-H4 transfected cells and not peripheral blood cells. Exact expression pattern of B7-H4 has not been fully characterized.