Please login to your online account to display your discounted pricing

B2, Tocris

Promotes inclusion formation in Huntington's and Parkinson's diseases

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material B2
Assay Percent Range >98%
Molecular Formula C20H17ClN4O3
Formula Weight 396.83
Synonym 5-[4-(4-Chlorobenzoyl)-1-piperazinyl]-8-nitroquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

285550

 
r & d systems
285550
50mg Each for $746.48

285510

 
r & d systems
285510
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 396.83
Quantity 50mg
Chemical Name or Material B2
Assay Percent Range >98%
Molecular Formula C20H17ClN4O3
CAS 115687-05-3
Synonym 5-[4-(4-Chlorobenzoyl)-1-piperazinyl]-8-nitroquinoline
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C