missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ B-Tribromoborazine, 96%

Manufacturer:  Alfa Aesar™ 4372306

 View more versions of this product

Catalog No. AA4372306



Specifications

Specifications

B-Tribromoborazine
White
5g
B3Br3H3N3
UN1759
PCLMMJIWMGUYKF-UHFFFAOYSA-N
2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
23278010
Powder
Moisture Sensitive
124°C to 128°C
13703-88-3
MFCD00058924
2,4,6-tribromoborazine
B1(NB(NB(N1)Br)Br)Br
317.187
317.2
96%

Chemical Identifiers

13703-88-3
317.187
PCLMMJIWMGUYKF-UHFFFAOYSA-N
23278010
B1(NB(NB(N1)Br)Br)Br
B3Br3H3N3
MFCD00058924
2,4,6-tribromoborazine
2,4,6-tribromo-1,3,5,2,4,6-triazatriborinane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Documents