BUCHI™ V-710 Vacuum Pump Systems
Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry
Manufacturer:  BUCHI™ 072000
 View more versions of this product
Catalog No. 05-402-136
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
PEEK, PTFE, Glass, FEP, FFKM
41 to 55dBA
104°F
100/240V
10° to 40°C
Laboratory and industrial Rotavapor™
3.1m3/Hour
Brushless DC
V-710
50/60Hz
37.8cm
14.87 in.
22.9 lb.
Power cable, sound absorber, vacuum tubing and connections and operating instructions
1600rpm
10.4kg
370w
100/240V 50/60Hz
CE, CSA
40°C
6.96 in.
11.68 in.
29.7cm