BUCHI™ V-710 Vacuum Pump Systems
Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry
Manufacturer:  BUCHI™ 072501
 View more versions of this product
Catalog No. 05-402-163
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
V-710
41 to 55dBA
Power cable, sound absorber, vacuum tubing and connections and operating instructions, V-802 LabVac vacuum controller, Secondary condenser with insulation
Brushless DC
6.96 in.
50/60Hz
370w
PEEK, PTFE, Glass, FEP, FFKM
3.1m3/Hour
40°C
29.7cm
100/240V
22.9 lb.
100/240V 50/60Hz
1600rpm
104°F
37.8cm
10.4kg
10° to 40°C
Laboratory and industrial Rotavapor™
CE, CSA
11.68 in.
14.87 in.