BUCHI™ V-710 Vacuum Pump Systems
Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry
Manufacturer:  BUCHI™ 072511
 View more versions of this product
Catalog No. 05-402-166
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
PEEK, PTFE, Glass, FEP, FFKM
22.9 lb.
104°F
100/240V 50/60Hz
Brushless DC
37.8cm
V-710
10° to 40°C
Laboratory and industrial Rotavapor™
Power cable, sound absorber, vacuum tubing and connections and operating instructions, V-802 LabVac vacuum controller, woulff bottle, secondary condenser with insulation
29.7cm
14.87 in.
41 to 55dBA
3.1m3/Hour
CE, CSA
1600rpm
100/240V
11.68 in.
10.4kg
370w
40°C
6.96 in.
50/60Hz