Learn More
missing translation for 'pleaseLogIn'
BUCHI V-710 Vacuum Pump Systems
Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry
Manufacturer:  BUCHI 072012
 View more versions of this product
Catalog No. 05-402-141
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: All V-700 Vacuum Pumps come with a power cable, sound absorber, vacuum tubing and connections and operating instructions. See table for packages with additional components.
Description
Description
Order Info
All models listed are dual voltage: operate on 120V 50/60Hz or 240V 50/60Hz.
Specifications
Specifications
Brushless DC
Power cable, sound absorber, vacuum tubing and connections and operating instructions, woulff bottle, secondary cold trap
CE, CSA
PEEK, PTFE, Glass, FEP, FFKM
41 to 55dBA
22.9 lb.
104°F
10°C
Laboratory and industrial Rotavapor™
11.68 in.
14.87 in.
6.96 in.
100/240V
1600rpm
100/240V 50/60Hz
V-710
370w
10.4kg
40°C
3.1m³/hr.
50/60Hz
29.7cm
37.8cm
Documents