BUCHI™ V-710 Vacuum Pump Systems
Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry
Manufacturer:  BUCHI™ 072010
 View more versions of this product
Catalog No. 05-402-139
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
100/240V 50/60Hz
Laboratory and industrial Rotavapor™
V-710
10° to 40°C
22.9 lb.
Brushless DC
50/60Hz
29.7cm
14.87 in.
Power cable, sound absorber, vacuum tubing and connections and operating instructions, woulff bottle
40°C
CE, CSA
3.1m3/Hour
PEEK, PTFE, Glass, FEP, FFKM
41 to 55dBA
370w
10.4kg
100/240V
11.68 in.
6.96 in.
37.8cm
104°F
1600rpm