Please login to your online account to display your discounted pricing

Alfa Aesar™ B-9430


Quantity 0.5mg

Manufacturer: alfa aesar™  J66729EXD

 View more versions of this product

Catalog No. AAJ66729EXD

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 1338.55
Chemical Name or Material B-9430
MDL Number MFCD08705408
Molecular Formula C64H95N19O13
Physical Form Solid
CAS 180981-09-3
Color White to off-white
Solubility Information Soluble in water at 1mg/ml
Storage Note 1 -30° to -10°C
Recommended Storage −30° to −10°C
Quantity 0.5mg
Sensitivity Air & Light Sensitive
Synonym D-Arg-Arg-Pro-Hyp-Gly-Igl-Ser-D-Igl-Oic-Arg; D-Arg-[Hyp3, Igl5, D-Igl7, Oic8]-Bradykinin trifluoroacetate salt

Specifications

Chemical Name or Material B-9430
CAS 180981-09-3
Molecular Formula C64H95N19O13
Formula Weight 1338.55
MDL Number MFCD08705408
Synonym D-Arg-Arg-Pro-Hyp-Gly-Igl-Ser-D-Igl-Oic-Arg; D-Arg-[Hyp3, Igl5, D-Igl7, Oic8]-Bradykinin trifluoroacetate salt
Accessories

−30° to −10°C

-30° to -10°C

SDS & Safety Documents