Learn More
Azure I, Thermo Scientific Chemicals
$73.01 - $765.38
Chemical Identifiers
CAS | 531-55-5 |
---|---|
Molecular Formula | C15H16ClN3S |
Molecular Weight (g/mol) | 305.824 |
MDL Number | MFCD00011935 |
InChI Key | DNDJEIWCTMMZBX-UHFFFAOYSA-N |
Synonym | azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech |
PubChem CID | 68275 |
IUPAC Name | dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride |
SMILES | CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1750806
|
Thermo Scientific Chemicals
A1750806 |
5 g |
Each for $73.01
|
|
|||||
AAA1750814
|
Thermo Scientific Chemicals
A1750814 |
25 g |
Each for $223.19
|
|
|||||
AAA1750822
|
Thermo Scientific Chemicals
A1750822 |
100 g |
Each for $765.38
|
|
|||||
Description
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.
Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.
Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
Chemical Identifiers
531-55-5 | |
305.824 | |
DNDJEIWCTMMZBX-UHFFFAOYSA-N | |
68275 | |
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-] |
C15H16ClN3S | |
MFCD00011935 | |
azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech | |
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride |
Specifications
531-55-5 | |
Odorless | |
MFCD00011935 | |
3922630 | |
azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech | |
DNDJEIWCTMMZBX-UHFFFAOYSA-N | |
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride | |
68275 | |
Azure I |
∼205°C (decomposition) | |
C15H16ClN3S | |
5 g | |
14,6058 | |
Soluble in 1mg/ml water. Sparingly soluble in alcohol. | |
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-] | |
305.824 | |
305.83 |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 208-511-2
RTECSNumber : SO5550000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only