Please login to your online account to display your discounted pricing

Azure A Eosinate, pure, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  190600250

 View more versions of this product

Catalog No. AC190600250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 1158.59
Quantity 25g
Color Green
EINECS Number 263-491-2
Purity Grade Pure
Packaging Glass bottle
Molecular Formula C20H6Br4O5·2C14H14N3S
MDL Number MFCD00012112
Chemical Name or Material Azure A Eosinate
CAS 62298-43-5
Infrared Spectrum Authentic
UV Spectrometry Complies

Specifications

Chemical Name or Material Azure A Eosinate
CAS 62298-43-5
Purity Grade Pure
Molecular Formula C20H6Br4O5·2C14H14N3S
Formula Weight 1158.59
MDL Number MFCD00012112