Please login to your online account to display your discounted pricing

Azure A Eosinate, pure, ACROS Organics™

$112.82 - $112.82

Chemical Identifiers

Chemical Name or Material Azure A Eosinate
CAS 62298-43-5
Purity Grade Pure
Molecular Formula C20H6Br4O5·2C14H14N3S
Formula Weight 1158.59
MDL Number MFCD00012112
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190600250

  SDS
thermo fisher scientific
190600250
Glass bottle 25g Each for $112.82
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 1158.59
Quantity 25g
Color Green
EINECS Number 263-491-2
Purity Grade Pure
Chemical Name or Material Azure A Eosinate
Molecular Formula C20H6Br4O5·2C14H14N3S
MDL Number MFCD00012112
Packaging Glass bottle
CAS 62298-43-5
Infrared Spectrum Authentic
UV Spectrometry Complies