missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Azulene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 105181000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Azulene | |
275-51-4 | |
Authentic | |
Glass bottle | |
MFCD00003810 | |
05, II, 432 | |
cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep | |
CUFNKYGDVFVPHO-UHFFFAOYSA-N | |
azulene | |
9231 | |
128.17 |
99% | |
242.0°C | |
99% | |
C10H8 | |
100mg | |
15, 919 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and acetone | |
C1=CC=C2C=CC=C2C=C1 | |
128.17 | |
CHEBI:31249 | |
99% |
Chemical Identifiers
275-51-4 | |
128.17 | |
CUFNKYGDVFVPHO-UHFFFAOYSA-N | |
9231 | |
azulene |
C10H8 | |
MFCD00003810 | |
cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep | |
CHEBI:31249 | |
C1=CC=C2C=CC=C2C=C1 |
Safety and Handling
EINECSNumber : 205-993-6
RTECSNumber : CO4570000
TSCA : TSCA
RUO â Research Use Only