missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azomethine-H hydrate, Alfa Aesar

Reagent for boron determination

Manufacturer:  Alfa Aesar 1681014

 View more versions of this product

Catalog No. AA1681014



Specifications

Specifications

Azomethine-H
32266-60-7
YWRFBXPQBUKIHP-KTKRTIGZSA-N
4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid
5702781
Crystalline
hydrate
C17H13NO8S2
C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1
423.41
423.40 (Anhydrous)
25g

Chemical Identifiers

32266-60-7
423.41
5702781
C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1
C17H13NO8S2
YWRFBXPQBUKIHP-KTKRTIGZSA-N
4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid
SDS Documents