missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azomethine-H hydrate, Thermo Scientific Chemicals

Reagent for boron determination

Supplier:  Thermo Scientific Chemicals 01681009

 View more versions of this product

Catalog No. AA1681009



Description

Description

Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Azomethine-H
32266-60-7
MFCD00066536
DUCCKQSNXPFEGT-NVMNQCDNSA-N
4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid
5702781
Light sensitive
Powder
hydrate
C17H13NO8S2
Soluble in acetone,ammonium hydroxide,n-butanol and water.
OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
423.41
423.40 (anhydrous)
10 g

Chemical Identifiers

32266-60-7
423.41
DUCCKQSNXPFEGT-NVMNQCDNSA-N
OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
C17H13NO8S2
MFCD00066536
5702781
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 250-975-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only