Please login to your online account to display your discounted pricing

Azomethine-H hydrate, Alfa Aesar™

$64.65 - $64.65

Chemical Identifiers

Chemical Name or Material Azomethine-H hydrate
CAS 32266-60-7
Molecular Formula C17H13NO8S2·xH2O
Formula Weight 423.40 (Anhydrous)
MDL Number MFCD00066536
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1681004

  SDS
alfa aesar
1681004
Each for $64.65
Description & Specifications

Specifications

Formula Weight 423.40 (Anhydrous)
Quantity 2g
MDL Number MFCD00066536
Chemical Name or Material Azomethine-H hydrate
CAS 32266-60-7
Molecular Formula C17H13NO8S2·xH2O
Physical Form Crystalline