Please login to your online account to display your discounted pricing

Azodicarbonamide, 97%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC105165000

 View more versions of this product

Catalog No. AC105165000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 116.08
Quantity 500g
TSCA TSCA
EINECS Number 204-650-8
RTECS Number LQ1040000
Chemical Name or Material Azodicarbonamide
Name Note 97%
Health Hazard 1 Danger
Assay Percent Range 97%
Molecular Formula C2H4N4O2
Packaging Plastic bottle
Linear Formula H2NCON=NCONH2
MDL Number MFCD00007958
CAS 123-77-3
Synonym 1, 1'-Azobisformamide
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Azodicarbonamide
Name Note 97%
CAS 123-77-3
Assay Percent Range 97%
Molecular Formula C2H4N4O2
Linear Formula H2NCON=NCONH2
Formula Weight 116.08
MDL Number MFCD00007958
Synonym 1, 1'-Azobisformamide
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents