Please login to your online account to display your discounted pricing

Azodicarbonamide, 97%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC105160025

 View more versions of this product

Catalog No. AC105160025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 116.08
Quantity 2.5kg
RTECS Number LQ1040000
EINECS Number 204-650-8
TSCA TSCA
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00007958
Linear Formula H2NCON=NCONH2
Chemical Name or Material Azodicarbonamide
CAS 123-77-3
Molecular Formula C2H4N4O2
Packaging Plastic bottle
Synonym 1, 1'-Azobisformamide
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Specifications

Chemical Name or Material Azodicarbonamide
Name Note 97%
CAS 123-77-3
Assay Percent Range 97%
Molecular Formula C2H4N4O2
Linear Formula H2NCON=NCONH2
Formula Weight 116.08
MDL Number MFCD00007958
Synonym 1, 1'-Azobisformamide
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents