missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azocarmine G

$61.88 - $241.66

Chemical Identifiers

CAS 25641-18-3
Molecular Formula C28H18N3NaO6S2
Formula Weight 579.58
MDL Number MFCD00042002
Synonym C.I. 50085
Chemical Name or Material Azocarmine G
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1250706 SDS Alfa Aesar™
A1250706
5g Each for $61.88
AAA1250714 SDS Alfa Aesar™
A1250714
25g Each for $241.66
Specifications

Description

Applications
Azocarmine G is a stain used for microscopy.

Solubility
Soluble in water (1 mg/ml).

Notes
Store at room temperature with adequate ventilation.
Description & Specifications

Chemical Identifiers

25641-18-3
579.58
C.I. 50085
C28H18N3NaO6S2
MFCD00042002
Azocarmine G

Specifications

25641-18-3
579.58
C.I. 50085
Azocarmine G
C28H18N3NaO6S2
MFCD00042002
5g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA