missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Azlon™ Solvent Venting Wash Bottles

Manufacturer:  Dynalon™ 5064750001

 View more versions of this product

Catalog No. 03-405-9


 

Description

Description

Translucent low density polyethylene wash bottles feature unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. Wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.

Order Info

Bottles are available in 250 or 500mL capacities with five common solvent labels.

Compliance

All bottles have SPI recycling code stamped on base.

Specifications

Specifications

8 oz., 250mL
8 oz.
Polypropylene
Custom
LDPE
58.92mm
White
250mL
SPI recycling code stamped on base
Translucent
8.12 in.
2.32 in.
No
206.24mm
Documents